Learn More
Biomatik Corporation Human Tumor Necrosis Factor Ligand Superfamily, Member 13 (TNFSF13) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55031
The Human Tumor Necrosis Factor Ligand Superfamily, Member 13 (TNFSF13) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 46.875-3000 pg/mL. Sensitivity: 16.64 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O75888. NCBI Gene ID: 8741. Restriction: For Research Use Only.
Catalog No. 50-150-1608