Learn More
Biomatik Corporation Human Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55076
The Human Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O43915. NCBI Gene ID: 2277. Restriction: For Research Use Only.
Catalog No. 50-150-1632