Learn More
Biomatik Corporation Mouse Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55077
The Mouse Vascular Endothelial Growth Factor D (VEGFD) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.4 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P97946. NCBI Gene ID: 14205. Restriction: For Research Use Only.
Catalog No. 50-150-1633