Learn More
Biomatik Corporation Rat Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55086
The Rat Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O08775. NCBI Gene ID: 25589. Restriction: For Research Use Only.
Catalog No. 50-150-1638