Learn More
Biomatik Corporation Human A Disintegrin And Metalloprotease 17 (ADAM17) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU02045
The Human A Disintegrin And Metalloprotease 17 (ADAM17) ELISA Kit allows for quantitative determination of A Disintegrin And Metalloprotease 17 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78536. Cross Activity: No significant cross-reactivity or interference between A Disintegrin And Metalloprotease 17 and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.