Learn More
Biomatik Corporation Gallus Adiponectin (ADP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02152
The Gallus Adiponectin (ADP) ELISA Kit allows for quantitative determination of Adiponectin in Serum, Plasma and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 125-8000 pg/mL. Sensitivity: 57 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q60994. Cross Activity: No significant cross-reactivity or interference between Adiponectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1699