Learn More
Biomatik Corporation Human Adiponectin (ADP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02153
The Human Adiponectin (ADP) ELISA Kit allows for quantitative determination of Adiponectin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.069 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q15848. Cross Activity: No significant cross-reactivity or interference between Adiponectin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1700