Learn More
Biomatik Corporation Human AE Binding Protein 1 (AEBP1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02196
The Human AE Binding Protein 1 (AEBP1) ELISA Kit allows for quantitative determination of AE Binding Protein 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IUX7. Cross Activity: No significant cross-reactivity or interference between AE Binding Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1722