Learn More
Biomatik Corporation Human Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02222
The Human Aldehyde Dehydrogenase 1 Family, Member A1 (ALDH1A1) ELISA Kit allows for quantitative determination of Aldehyde Dehydrogenase 1 Family, Member A1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.63 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00352. Cross Activity: No significant cross-reactivity or interference between Aldehyde Dehydrogenase 1 Family, Member A1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1734