Learn More
Biomatik Corporation Mouse Alpha-2-Macroglobulin (a2M) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02279
The Mouse Alpha-2-Macroglobulin (a2M) ELISA Kit allows for quantitative determination of Alpha-2-Macroglobulin in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 3.70-300 ng/mL. Sensitivity: 1.51 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: Q61838. Cross Activity: No significant cross-reactivity or interference between Alpha-2-Macroglobulin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1760