Learn More
Biomatik Corporation Human Angiopoietin 2 (ANGPT2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02361
The Human Angiopoietin 2 (ANGPT2) ELISA Kit allows for quantitative determination of Angiopoietin 2 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 12.5-800 pg/mL. Sensitivity: 5.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15123. Cross Activity: No significant cross-reactivity or interference between Angiopoietin 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1790