Learn More
Biomatik Corporation Human Angiostatin (ANG) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02382
The Human Angiostatin (ANG) ELISA Kit allows for quantitative determination of Angiostatin in Serum, Plasma, Urine and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P00747. Cross Activity: No significant cross-reactivity or interference between Angiostatin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1799