Learn More
Biomatik Corporation Human B-Cell Activating Factor (BAFF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02664
The Human B-Cell Activating Factor (BAFF) ELISA Kit allows for quantitative determination of B-Cell Activating Factor in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 28 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y275. Cross Activity: No significant cross-reactivity or interference between B-Cell Activating Factor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1906