Learn More
Biomatik Corporation Rat B-Lymphocyte Chemoattractant 1 (BLC1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02730
The Rat B-Lymphocyte Chemoattractant 1 (BLC1) ELISA Kit allows for quantitative determination of B-Lymphocyte Chemoattractant 1 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5I0J6. Cross Activity: No significant cross-reactivity or interference between B-Lymphocyte Chemoattractant 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1927