Learn More
Biomatik Corporation Pig C Reactive Protein (CRP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02825
The Pig C Reactive Protein (CRP) ELISA Kit allows for quantitative determination of C Reactive Protein in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Sus scrofa; Porcine (Pig). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.31 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O19062. Cross Activity: No significant cross-reactivity or interference between C Reactive Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-1969