Learn More
Biomatik Corporation Rat Calcitonin Gene Related Peptide (CGRP) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU02858
The Rat Calcitonin Gene Related Peptide (CGRP) ELISA Kit allows for quantitative determination of Calcitonin Gene Related Peptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.94 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P10093. Cross Activity: No significant cross-reactivity or interference between Calcitonin Gene Related Peptide and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.