Learn More
Biomatik Corporation Human Carbohydrate Antigen 125 (CA125) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02908
The Human Carbohydrate Antigen 125 (CA125) ELISA Kit allows for quantitative determination of Carbohydrate Antigen 125 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100U/mL. Sensitivity: 0.58U/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8WXI7. Cross Activity: No significant cross-reactivity or interference between Carbohydrate Antigen 125 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2002