Learn More
Biomatik Corporation Human Cartilage Intermediate Layer Protein 2 (CILP2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU02972
The Human Cartilage Intermediate Layer Protein 2 (CILP2) ELISA Kit allows for quantitative determination of Cartilage Intermediate Layer Protein 2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.116 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q8IUL8. Cross Activity: No significant cross-reactivity or interference between Cartilage Intermediate Layer Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2021