Learn More
Biomatik Corporation Mouse Chitinase 1 (CHIT1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03126
The Mouse Chitinase 1 (CHIT1) ELISA Kit allows for quantitative determination of Chitinase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9D7Q1. Cross Activity: No significant cross-reactivity or interference between Chitinase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2092