Learn More
Biomatik Corporation Dog Cholecystokinin (CCK) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03131
The Dog Cholecystokinin (CCK) ELISA Kit allows for quantitative determination of Cholecystokinin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 12.35-1000 pg/mL. Sensitivity: 5.25 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: G1K283. Cross Activity: No significant cross-reactivity or interference between Cholecystokinin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2095