Learn More
Biomatik Corporation Human Chorionic Gonadotropin (CG) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03173
The Human Chorionic Gonadotropin (CG) ELISA Kit allows for quantitative determination of Chorionic Gonadotropin in Serum, Plasma, Urine, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.78-50 ng/mL. Sensitivity: 0.32 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P0DN86. Cross Activity: No significant cross-reactivity or interference between Chorionic Gonadotropin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2106