Learn More
Biomatik Corporation Human Coagulation Factor VII (F7) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03266
The Human Coagulation Factor VII (F7) ELISA Kit allows for quantitative determination of Coagulation Factor VII in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 9.4-600 pg/mL. Sensitivity: 3.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08709. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor VII and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2146