Learn More
Biomatik Corporation Human Coagulation Factor XI (F11) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03275
The Human Coagulation Factor XI (F11) ELISA Kit allows for quantitative determination of Coagulation Factor XI in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.227 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P03951. Cross Activity: No significant cross-reactivity or interference between Coagulation Factor XI and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2152