Learn More
Biomatik Corporation Rat Collagen Type I (COL1) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03299
The Rat Collagen Type I (COL1) ELISA Kit allows for quantitative determination of Collagen Type I in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.239 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. Cross Activity: No significant cross-reactivity or interference between Collagen Type I and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.