Learn More
Biomatik Corporation Human Collagen Type VI (COL6) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03332
The Human Collagen Type VI (COL6) ELISA Kit allows for quantitative determination of Collagen Type VI in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 ng/mL. Sensitivity: 2.8 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75794. Cross Activity: No significant cross-reactivity or interference between Collagen Type VI and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2175