Learn More
Biomatik Corporation Mouse Complement 1 Inhibitor (C1INH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03367
Encompass_Preferred
The Mouse Complement 1 Inhibitor (C1INH) ELISA Kit allows for quantitative determination of Complement 1 Inhibitor in Serum, Plasma and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.62-1000 ng/mL. Sensitivity: 5.9 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P97290. Cross Activity: No significant cross-reactivity or interference between Complement 1 Inhibitor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2190