Learn More
Biomatik Corporation Rat Complement Component 3a (C3a) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03390
The Rat Complement Component 3a (C3a) ELISA Kit allows for quantitative determination of Complement Component 3a in Serum, Plasma and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.81 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01026. Cross Activity: No significant cross-reactivity or interference between Complement Component 3a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2200