Learn More
Biomatik Corporation Mouse Complement Component 5a (C5a) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKU03407
The Mouse Complement Component 5a (C5a) ELISA Kit allows for quantitative determination of Complement Component 5a in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.57 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P06684. Cross Activity: No significant cross-reactivity or interference between Complement Component 5a and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2212