Learn More
Biomatik Corporation Human Complement Factor H Related Protein 3 (CFHR3) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03427
The Human Complement Factor H Related Protein 3 (CFHR3) ELISA Kit allows for quantitative determination of Complement Factor H Related Protein 3 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.075 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q02985. Cross Activity: No significant cross-reactivity or interference between Complement Factor H Related Protein 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2225