Learn More
Biomatik Corporation Mouse Cross Linked C-Telopeptide of Type I Collagen (CTXI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03503
Encompass_Preferred
The Mouse Cross Linked C-Telopeptide of Type I Collagen (CTXI) ELISA Kit allows for quantitative determination of Cross Linked C-Telopeptide of Type I Collagen in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 123.5-10,000 pg/mL. Sensitivity: 49.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Cross Linked C-Telopeptide of Type I Collagen and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2263