Learn More
Biomatik Corporation Human Cyclin Dependent Kinase Inhibitor 2A (CDKN2A) ELISA Kit, 96 tests
Supplier: Biomatik Corporation EKU03545
The Human Cyclin Dependent Kinase Inhibitor 2A (CDKN2A) ELISA Kit allows for quantitative determination of Cyclin Dependent Kinase Inhibitor 2A in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.262 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42771. Cross Activity: No significant cross-reactivity or interference between Cyclin Dependent Kinase Inhibitor 2A and analogues was observed. Restriction: For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.