Learn More
Biomatik Corporation Mouse Cystatin A (CSTA) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03568
The Mouse Cystatin A (CSTA) ELISA Kit allows for quantitative determination of Cystatin A in Serum, Plasma, Saliva, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 62.5-4000 pg/mL. Sensitivity: 25.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P56567. Cross Activity: No significant cross-reactivity or interference between Cystatin A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2290