Learn More
Biomatik Corporation Human Cysteine Rich Protein 1, Intestinal (CRIP1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03572
The Human Cysteine Rich Protein 1, Intestinal (CRIP1) ELISA Kit allows for quantitative determination of Cysteine Rich Protein 1, Intestinal in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P50238. Cross Activity: No significant cross-reactivity or interference between Cysteine Rich Protein 1, Intestinal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2291