Learn More
Biomatik Corporation Mouse Cysteine Rich Protein, Angiogenic Inducer 61 (CYR61) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03574
The Mouse Cysteine Rich Protein, Angiogenic Inducer 61 (CYR61) ELISA Kit allows for quantitative determination of Cysteine Rich Protein, Angiogenic Inducer 61 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25-400 pg/mL. Sensitivity: 2.36 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P18406. Cross Activity: No significant cross-reactivity or interference between Cysteine Rich Protein, Angiogenic Inducer 61 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2292