Learn More
Biomatik Corporation Human Defensin Beta 103A (DEFb103A) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03661
The Human Defensin Beta 103A (DEFb103A) ELISA Kit allows for quantitative determination of Defensin Beta 103A in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 46.88-3000 pg/mL. Sensitivity: 19.54 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P81534. Cross Activity: No significant cross-reactivity or interference between Defensin Beta 103A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2319