Learn More
Biomatik Corporation Human Defensin Beta 126 (DEFb126) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03665
Encompass_Preferred
The Human Defensin Beta 126 (DEFb126) ELISA Kit allows for quantitative determination of Defensin Beta 126 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BYW3. Cross Activity: No significant cross-reactivity or interference between Defensin Beta 126 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2320