Learn More
Biomatik Corporation Human Deiodinase, Iodothyronine, Type II (DIO2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03674
The Human Deiodinase, Iodothyronine, Type II (DIO2) ELISA Kit allows for quantitative determination of Deiodinase, Iodothyronine, Type II in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92813. Cross Activity: No significant cross-reactivity or interference between Deiodinase, Iodothyronine, Type II and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2324