Learn More
Biomatik Corporation Mouse Delta Like 1 Homolog (dLK1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03681
The Mouse Delta Like 1 Homolog (dLK1) ELISA Kit allows for quantitative determination of Delta Like 1 Homolog in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q09163. Cross Activity: No significant cross-reactivity or interference between Delta Like 1 Homolog and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2327