Learn More
Biomatik Corporation Human DNA Methyltransferase 3A (DNMT3A) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU03770
The Human DNA Methyltransferase 3A (DNMT3A) ELISA Kit allows for quantitative determination of DNA Methyltransferase 3A in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9Y6K1. Cross Activity: No significant cross-reactivity or interference between DNA Methyltransferase 3A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2362