Learn More
Biomatik Corporation Mouse Fatty Acid Binding Protein 5, Epidermal (FABP5) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04042
The Mouse Fatty Acid Binding Protein 5, Epidermal (FABP5) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 5, Epidermal in Tissue homogenates, Breast milk and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q05816. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 5, Epidermal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2451