Learn More
Biomatik Corporation Rat Fatty Acid Transport Protein 1 (FATP1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04056
The Rat Fatty Acid Transport Protein 1 (FATP1) ELISA Kit allows for quantitative determination of Fatty Acid Transport Protein 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P97849. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Transport Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2459