Learn More
Biomatik Corporation Human Fibrinogen Alpha (FGa) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04093
The Human Fibrinogen Alpha (FGa) ELISA Kit allows for quantitative determination of Fibrinogen Alpha in Plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 ng/mL. Sensitivity: 6.5 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02671. Cross Activity: No significant cross-reactivity or interference between Fibrinogen Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2477