Learn More
Biomatik Corporation Human Fibrinogen Like Protein 2 (FGL2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04109
The Human Fibrinogen Like Protein 2 (FGL2) ELISA Kit allows for quantitative determination of Fibrinogen Like Protein 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.19 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14314. Cross Activity: No significant cross-reactivity or interference between Fibrinogen Like Protein 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2483