Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04154
The Mouse Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 2, Basic in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.41 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. UniProt ID: P15655. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 2, Basic and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2493