Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 7 (FGF7) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04178
The Mouse Fibroblast Growth Factor 7 (FGF7) ELISA Kit allows for quantitative determination of Fibroblast Growth Factor 7 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8-500 pg/mL. Sensitivity: 2.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P36363. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 7 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2504