Learn More
Biomatik Corporation Human Fibulin 4 (FBLN4) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04213
The Human Fibulin 4 (FBLN4) ELISA Kit allows for quantitative determination of Fibulin 4 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.12 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O95967. Cross Activity: No significant cross-reactivity or interference between Fibulin 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2519