Learn More
Biomatik Corporation Human FK506 Binding Protein 12 Rapamycin Associated Protein (FRAP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04228
The Human FK506 Binding Protein 12 Rapamycin Associated Protein (FRAP) ELISA Kit allows for quantitative determination of FK506 Binding Protein 12 Rapamycin Associated Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.114 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42345. Cross Activity: No significant cross-reactivity or interference between FK506 Binding Protein 12 Rapamycin Associated Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2526