Learn More
Biomatik Corporation Rat Galectin 3 (GAL3) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04321
The Rat Galectin 3 (GAL3) ELISA Kit allows for quantitative determination of Galectin 3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus (Rat). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08699. Cross Activity: No significant cross-reactivity or interference between Galectin 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2555