Learn More
Biomatik Corporation Human Galectin 8 (GAL8) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04325
Encompass_Preferred
The Human Galectin 8 (GAL8) ELISA Kit allows for quantitative determination of Galectin 8 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00214. Cross Activity: No significant cross-reactivity or interference between Galectin 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2556