Learn More
Biomatik Corporation Mouse Galectin 8 (GAL8) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKU04326
The Mouse Galectin 8 (GAL8) ELISA Kit allows for quantitative determination of Galectin 8 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Mus musculus (Mouse). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9JL15. Cross Activity: No significant cross-reactivity or interference between Galectin 8 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2557